BalanCD™ CHO Feed 1
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91127-1L 94119-10L
77975-320EA 313.66 CAD
77975-320 77975-391
BalanCD™ CHO Feed 1
Cell Culture Supplements and Additives
BalanCD CHO Feed 1 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode. This formulation was developed using FUJIFILM Irvine Scientific’s rational culture media design strategy to achieve enhanced performance of growth and production in fed-batch culture. BalanCD CHO Feed 1 was designed to be used in conjunction with BalanCD CHO Growth A and can work with a wide range of growth media for optimal performance in fed-batch cultures.
  • All BalanCD CHO growth media and feeds are formulated for industrial applications and transition smoothly into commercial production
  • Purpose-built formulations for fed batch bioprocesses
  • Rational culture media design approach ensures scalability to commercial production
  • BalanCD CHO feeds are formulated using chemically-defined components in high concentrations based on typical CHO cell requirements
Packaging: Store at 2 to 8 °C. Shelf life: One year from date of manufacture.