BalanCD™ Gal Supplement
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91175-100ML 91175-1L
77975-342EA 167.4 CAD
77975-342 77975-343
BalanCD™ Gal Supplement
Cell Culture Supplements and Additives
The ability to produce high product titers and excellent protein quality is critical for the generation of effective therapeutic biomolecules. Glycosylation modifications, including galactosylation, help to achieve this critical quality aspect by enhancing protein characteristics and antibody efficacy. BalanCD™ Gal Supplement delivers increased galactosylation, through the addition of N-linked glycan moieties, while supporting an efficient, streamlined workflow.
  • Compatible with any basal growth medium for a wide range of cell lines
  • Simplified, scalable protocol
  • Enhanced formula requires a lower concentration to achieve desired outcome - contributing to improved efficiency and streamlined workflow
  • Simple, single feeding schedule
  • Chemically-defined, animal component-free, and cGMP manufactured to support commercial applications
  • Designed to incorporate seamlessly into the BalanCD CHO Media
  • Platform for efficient growth and production
Packaging:
Store at 15 to 30 °C
Shelf life: 24 months from date of manufacture