BalanCD™ CHO Feed 3
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
94118-10L 99471-1L
77975-390EA 2661.98 CAD
77975-390 77975-401
BalanCD™ CHO Feed 3
Cell Culture Supplements and Additives
BalanCD™ CHO Feed 3 is a chemically-defined feed supplement of non-animal origin. Using Irvine Scientific’s Rational Culture Media Design® approach, this formulation has been developed to achieve high growth kinetics and product yield in fed-batch processes. The feed supplement contains balanced concentrations of glucose, amino acids, vitamins and trace elements to provide nutrients to leaner batch media that are needed to boost the productivity of the cell culture system in fed-batch.
  • Flexible, scalable, and expertly formulated to maximize productivity, growth, and viability of CHO cell lines
  • Designed to nurture and advance every step of development
  • BalanCD CHO Feeds are formulated using chemically-defined components in high concentrations based on typical CHO cell requirements
Packaging:
Store at 2 to 8 °C
Shelf life: One year from date of manufacture