HBSS 1X-Hanks' Balanced Salt Solutions
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
9220-100ML
9220-500ML
9228-100ML
9228-500ML
77975-364EA
22.67
CAD
77975-364
77975-365
77975-366
77975-367
HBSS 1X-Hanks' Balanced Salt Solutions
Buffers
Cell Culture Buffers
HBSS 1X Hanks' Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions. HBSS is suitable for use in sealed culture flasks incubated in air.
- Calcium and magnesium salts
- Phenol red
- Sodium bicarbonate
Packaging: Store at 15 to 30 °C and protect from light. Shelf life: 24 months from date of manufacture.