PRIME-XV MSC Expansion XSFM
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
Human mesenchymal stromal cell (MSC) expansion xeno-free, serum-free medium.
PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expansion of purified or enriched human mesenchymal stromal cells (MSCs) under serum-free and xeno-free culture conditions. This medium is ready to use. It may also be used with additional cytokine/growth factors for desired application. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.
Certifications: FDA, Federal, and State registered - cGMP-compliant manufacture EN ISO 13485:2016 certified MDSAP certified Extensive QC testing including functionality, sterility (USP <71>), endotoxin (USP <85>), and mycoplasma (USP <63>) Drug Master Files (DMFs) filed with the FDA – available upon request.