BalanCD™ CHO Feed 4
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
94134-10L 94134-1L
77975-396EA 3326.67 CAD
77975-396 77975-397
BalanCD™ CHO Feed 4
Cell Culture Supplements and Additives
Maximize productivity in large-scale, fed-batch cultures.
  • Enriched feed improves productivity and cell growth
  • Advanced, concentrated formula achieves protein quality and provides flexibility to meet specific user needs
  • Simple three-step, pH neutral, hydration protocol saves labor and facilitates a more robust process
  • Fulfills regulatory requirements: chemically defined, animal-component free
BalanCD CHO Feed 4 is a nutrient-rich supplement designed to enable high-performing CHO cell lines achieve their full production potential while maintaining protein quality and functionality. Optimized for use with BalanCD CHO Growth A medium, BalanCD CHO Feed 4 is part of the scalable BalanCD CHO media platform used to produce high yields of high quality recombinant proteins.
Packaging: Store at 2 to 8 °C. Shelf life: Three years from date of manufacture.