Gold, nanoparticles, 40 nm
Supplier: Abcam
AB269928-10ML AB269928-100ML
ABCA_AB269928-10MLEA 594.08 CAD
ABCA_AB269928-10ML ABCA_AB269928-100M
Gold, nanoparticles, 40 nm
Gold
40nm Gold Nanoparticles (1 OD) is an aqueous suspension of spherical metallic nanoparticles.