Anti-Syntaxin 1a Rabbit Monoclonal Antibody [clone: EPR23457-15]

Supplier: Abcam
AB272736-100UL AB272736-1ML
ABCA_AB272736-100UEA 1088.62 CAD
ABCA_AB272736-100U ABCA_AB272736-1ML
Anti-Syntaxin 1a Rabbit Monoclonal Antibody [clone: EPR23457-15]
Antibodies
Rabbit monoclonal [EPR23457-15] to Syntaxin 1a.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Syntaxin 1a
Clonality: Monoclonal
Clone: EPR23457-15
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR