Anti-Thiophosphate ester Rabbit Monoclonal Antibody [clone: 51-8]

Supplier: Abcam
AB239919-1MG AB239919-100UG
ABCA_AB239919-1MGEA 16158.66 CAD
ABCA_AB239919-1MG ABCA_AB239919-100U
Anti-Thiophosphate ester Rabbit Monoclonal Antibody [clone: 51-8]
Antibodies
Rabbit monoclonal [51-8] to Thiophosphate ester - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Thiophosphate ester
Clonality: Monoclonal
Clone: 51-8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR