Anti-Smad2+Smad3 Mouse Monoclonal Antibody [clone: 18-Smad2-3]

Supplier: Abcam
AB305325-1MG AB305325-10UG AB305325-20UG AB305325-100UG
ABCA_AB305325-1MGEA 11287.52 CAD
ABCA_AB305325-1MG ABCA_AB305325-10UG ABCA_AB305325-20UG ABCA_AB305325-100U
Anti-Smad2+Smad3 Mouse Monoclonal Antibody [clone: 18-Smad2-3]
Antibodies
Mouse monoclonal [18/Smad2/3] to Smad2 + Smad3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Smad2+Smad3
Clonality: Monoclonal
Clone: 18-Smad2-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR