Anti-Uridine Phosphorylase 1 Rabbit Monoclonal Antibody [clone: EPR7680]

Supplier: Abcam
AB128854-100UL AB128854-1ML
ABCA_AB128854-100UEA 881.64 CAD
ABCA_AB128854-100U ABCA_AB128854-1ML
Anti-Uridine Phosphorylase 1 Rabbit Monoclonal Antibody [clone: EPR7680]
Antibodies
Rabbit monoclonal [EPR7680] to Uridine Phosphorylase 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Uridine Phosphorylase 1
Clonality: Monoclonal
Clone: EPR7680
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR