Anti-CD204 Rabbit Monoclonal Antibody [clone: 4] PE (Phycoerythrin)

Supplier: Abcam
AB275707-25TEST AB275707-100TEST
ABCA_AB275707-25TEEA 385.52 CAD
ABCA_AB275707-25TE ABCA_AB275707-100T
Anti-CD204 Rabbit Monoclonal Antibody [clone: 4] PE (Phycoerythrin)
Antibodies
PE Rabbit monoclonal [004] to CD204.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD204
Clonality: Monoclonal
Clone: 4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR