Anti-Carbonic Anhydrase 9/CA9 Rabbit Monoclonal Antibody [clone: 53] PE (Phycoerythrin)

Supplier: Abcam
AB275578-100TEST AB275578-25TEST
ABCA_AB275578-100TEA 1099.68 CAD
ABCA_AB275578-100T ABCA_AB275578-25TE
Anti-Carbonic Anhydrase 9/CA9 Rabbit Monoclonal Antibody [clone: 53] PE (Phycoerythrin)
Antibodies
PE Rabbit monoclonal [053] to Carbonic Anhydrase 9/CA9.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 9/CA9
Clonality: Monoclonal
Clone: 53
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR