Anti-Ki67 Mouse Monoclonal Antibody [clone: 37C7-12]

Supplier: Abcam
AB245113-100UG AB245113-1MG
ABCA_AB245113-100UEA 1257.68 CAD
ABCA_AB245113-100U ABCA_AB245113-1MG
Anti-Ki67 Mouse Monoclonal Antibody [clone: 37C7-12]
Antibodies
Mouse monoclonal [37C7-12] to Ki67.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ki67
Clonality: Monoclonal
Clone: 37C7-12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR