Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: SP53]

Supplier: Abcam
AB119695-1ML AB119695-20UL AB119695-100UL AB119695-500UL
ABCA_AB119695-1MLEA 1703.24 CAD
ABCA_AB119695-1ML ABCA_AB119695-20UL ABCA_AB119695-100U ABCA_AB119695-500U
Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: SP53]
Antibodies
Rabbit monoclonal [SP53] to Cytokeratin 14.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 14
Clonality: Monoclonal
Clone: SP53
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR