Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: YCA-R20133-107 H3L2]

Supplier: Abcam
AB288610-100UG AB288610-1MG
ABCA_AB288610-100UEA 1564.2 CAD
ABCA_AB288610-100U ABCA_AB288610-1MG
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: YCA-R20133-107 H3L2]
Antibodies
Rabbit monoclonal [YCA-R20133-107 H3L2] to Antithrombin III/ATIII - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Antithrombin III/ATIII
Clonality: Monoclonal
Clone: YCA-R20133-107 H3L2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR