Anti-Cytokeratin 20 Rabbit Monoclonal Antibody [clone: SP33]

Supplier: Abcam
AB64090-1ML AB64090-100UL AB64090-500UL
ABCA_AB64090-1MLEA 1951.3 CAD
ABCA_AB64090-1ML ABCA_AB64090-100UL ABCA_AB64090-500UL
Anti-Cytokeratin 20 Rabbit Monoclonal Antibody [clone: SP33]
Antibodies
Rabbit monoclonal [SP33] to Cytokeratin 20.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 20
Clonality: Monoclonal
Clone: SP33
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR