Anti-SLP76 (phospho Y128) Rabbit Monoclonal Antibody [clone: SLP76Y128-3F8] (APC)

Supplier: Abcam
AB278579-100TEST AB278579-10TEST
ABCA_AB278579-100TEA 1545.24 CAD
ABCA_AB278579-100T ABCA_AB278579-10TE
Anti-SLP76 (phospho Y128) Rabbit Monoclonal Antibody [clone: SLP76Y128-3F8] (APC)
Antibodies
APC Rabbit monoclonal [SLP76Y128-3F8] to SLP76 (phospho Y128).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SLP76
Clonality: Monoclonal
Clone: SLP76Y128-3F8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR