Anti-HC-II Rabbit Monoclonal Antibody [clone: EPR25618-1]

Supplier: Abcam
AB289971-100UL AB289971-1ML AB289971-20UL
ABCA_AB289971-100UEA 1297.18 CAD
ABCA_AB289971-100U ABCA_AB289971-1ML ABCA_AB289971-20UL
Anti-HC-II Rabbit Monoclonal Antibody [clone: EPR25618-1]
Antibodies
Rabbit monoclonal [EPR25618-1] to HC-II.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HC-II
Clonality: Monoclonal
Clone: EPR25618-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR