Anti-PKC (phospho T514) Rabbit Monoclonal Antibody [clone: PKCgT514-PF4] (PE)

Supplier: Abcam
AB278661-10TEST AB278661-100TEST
ABCA_AB278661-10TEEA 524.56 CAD
ABCA_AB278661-10TE ABCA_AB278661-100T
Anti-PKC (phospho T514) Rabbit Monoclonal Antibody [clone: PKCgT514-PF4] (PE)
Antibodies
PE Rabbit monoclonal [PKCgT514-PF4] to PKC (phospho T514).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PKC
Clonality: Monoclonal
Clone: PKCgT514-PF4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR