Anti-non-muscle Myosin IIB/MYH10 Goat Chimera Monoclonal Antibody [clone: EPR22564-23]

Supplier: Abcam
AB300649-100UG AB300649-1MG
ABCA_AB300649-100UEA 1791.72 CAD
ABCA_AB300649-100U ABCA_AB300649-1MG
Anti-non-muscle Myosin IIB/MYH10 Goat Chimera Monoclonal Antibody [clone: EPR22564-23]
Antibodies
Goat Chimera monoclonal [EPR22564-23] to non-muscle Myosin IIB/MYH10 - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: non-muscle Myosin IIB/MYH10
Clonality: Monoclonal
Clone: EPR22564-23
Conjugation:
Epitope:
Host: Goat Chimera
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR