Anti-GFP Rat Monoclonal Antibody [clone: 3H9]

Supplier: Abcam
AB252881-100UG AB252881-1MG AB252881-20UG
ABCA_AB252881-100UEA 791.58 CAD
ABCA_AB252881-100U ABCA_AB252881-1MG ABCA_AB252881-20UG
Anti-GFP Rat Monoclonal Antibody [clone: 3H9]
Antibodies
Rat monoclonal [3H9] to GFP.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: 3H9
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR