Anti-Tyrosine Hydroxylase Rabbit Monoclonal Antibody [clone: EP1532Y] (Alexa Fluor® 555)

Supplier: Abcam
AB305365-100UL AB305365-10UL
ABCA_AB305365-100UEA 1148.66 CAD
ABCA_AB305365-100U ABCA_AB305365-10UL
Anti-Tyrosine Hydroxylase Rabbit Monoclonal Antibody [clone: EP1532Y] (Alexa Fluor® 555)
Antibodies
Alexa Fluor® 555 Rabbit monoclonal [EP1532Y] to Tyrosine Hydroxylase - Neuronal Marker.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR