Anti-alpha Tubulin (acetyl K40) Rat Monoclonal Antibody [clone: EPR16772]

Supplier: Abcam
AB289863-100UL AB289863-20UL AB289863-10UL AB289863-1ML
ABCA_AB289863-100UEA 791.58 CAD
ABCA_AB289863-100U ABCA_AB289863-20UL ABCA_AB289863-10UL ABCA_AB289863-1ML
Anti-alpha Tubulin (acetyl K40) Rat Monoclonal Antibody [clone: EPR16772]
Antibodies
Rat monoclonal [EPR16772] to alpha Tubulin (acetyl K40) - Chimeric.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha Tubulin
Clonality: Monoclonal
Clone: EPR16772
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR