Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EP5372]

Supplier: Abcam
AB244656-1MG AB244656-100UG
ABCA_AB244656-1MGEA 15534.56 CAD
ABCA_AB244656-1MG ABCA_AB244656-100U
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EP5372]
Antibodies
Rabbit monoclonal [EP5372] to Antithrombin III/ATIII - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Antithrombin III/ATIII
Clonality: Monoclonal
Clone: EP5372
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR