Anti-smooth muscle Myosin heavy chain 11 Rabbit Monoclonal Antibody [clone: SP314]

Supplier: Abcam
AB224804-1ML AB224804-100UL
ABCA_AB224804-1MLEA 1515.22 CAD
ABCA_AB224804-1ML ABCA_AB224804-100U
Anti-smooth muscle Myosin heavy chain 11 Rabbit Monoclonal Antibody [clone: SP314]
Antibodies
Rabbit monoclonal [SP314] to smooth muscle Myosin heavy chain 11 - N-terminal.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: smooth muscle Myosin heavy chain 11
Clonality: Monoclonal
Clone: SP314
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR