Anti-Histone H3 (acetyl K9) Rabbit Monoclonal Antibody [clone: EPR21238]

Supplier: Abcam
AB241150-1ML AB241150-100UL
ABCA_AB241150-1MLEA 9911.34 CAD
ABCA_AB241150-1ML ABCA_AB241150-100U
Anti-Histone H3 (acetyl K9) Rabbit Monoclonal Antibody [clone: EPR21238]
Antibodies
Rabbit monoclonal [EPR21238] to Histone H3 (acetyl K9).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR21238
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR