Anti-hnRNP A1 (citrulline R97) Rabbit Monoclonal Antibody [clone: EPR20176]

Supplier: Abcam
AB208028-100UL AB208028-1ML
ABCA_AB208028-100UEA 1079.14 CAD
ABCA_AB208028-100U ABCA_AB208028-1ML
Anti-hnRNP A1 (citrulline R97) Rabbit Monoclonal Antibody [clone: EPR20176]
Antibodies
Rabbit monoclonal [EPR20176] to hnRNP A1 (citrulline R97).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: hnRNP A1
Clonality: Monoclonal
Clone: EPR20176
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR