Anti-Inhibin alpha Rabbit Monoclonal Antibody [clone: EPY2782]

Supplier: Abcam
AB81322-100UL AB81322-1ML
ABCA_AB81322-100ULEA 1060.18 CAD
ABCA_AB81322-100UL ABCA_AB81322-1ML
Anti-Inhibin alpha Rabbit Monoclonal Antibody [clone: EPY2782]
Antibodies
Rabbit monoclonal [EPY2782] to Inhibin alpha.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Inhibin alpha
Clonality: Monoclonal
Clone: EPY2782
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR