Anti-Histone H2B (acetyl K12 + K12 + K16 + K23 + K23 + K23 + K23 + K23 + K5 + K7) Rabbit Monoclonal Antibody [clone: IGX4228R-1]

Supplier: Abcam
AB213225-100UL AB213225-1ML
ABCA_AB213225-100UEA 960.64 CAD
ABCA_AB213225-100U ABCA_AB213225-1ML
Anti-Histone H2B (acetyl K12 + K12 + K16 + K23 + K23 + K23 + K23 + K23 + K5 + K7) Rabbit Monoclonal Antibody [clone: IGX4228R-1]
Antibodies
Rabbit monoclonal [IGX4228R-1] to Histone H2B.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: IGX4228R-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human;Mouse;;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR