Anti-N Cadherin Mouse Monoclonal Antibody [clone: 32/N-Cadherin]

Supplier: Abcam
AB280375-20UG AB280375-1MG AB280375-100UG
ABCA_AB280375-20UGEA 455.04 CAD
ABCA_AB280375-20UG ABCA_AB280375-1MG ABCA_AB280375-100U
Anti-N Cadherin Mouse Monoclonal Antibody [clone: 32/N-Cadherin]
Antibodies
Mouse monoclonal [32/N-Cadherin] to N Cadherin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: N Cadherin
Clonality: Monoclonal
Clone: 32/N-Cadherin
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR