Anti-IL1 Receptor I/IL-1R-1 Rabbit Monoclonal Antibody [clone: 191] (APC)

Supplier: Abcam
AB279599-100TEST AB279599-25TEST
ABCA_AB279599-100TEA 1069.66 CAD
ABCA_AB279599-100T ABCA_AB279599-25TE
Anti-IL1 Receptor I/IL-1R-1 Rabbit Monoclonal Antibody [clone: 191] (APC)
Antibodies
APC Rabbit monoclonal [191] to IL1 Receptor I/IL-1R-1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL1 Receptor I/IL-1R-1
Clonality: Monoclonal
Clone: 191
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR