Anti-Strep-tag II Rabbit Monoclonal Antibody [clone: EPR12666]

Supplier: Abcam
AB180957-100UL AB180957-1ML
ABCA_AB180957-100UEA 791.58 CAD
ABCA_AB180957-100U ABCA_AB180957-1ML
Anti-Strep-tag II Rabbit Monoclonal Antibody [clone: EPR12666]
Antibodies
Rabbit monoclonal [EPR12666] to Strep-tag II.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Strep-tag II
Clonality: Monoclonal
Clone: EPR12666
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR