Anti-Collagen IV Rabbit Recombinant Multiclonal Antibody [clone: RM1065]

Supplier: Abcam
AB315250-1ML AB315250-100UL AB315250-20UL
ABCA_AB315250-1MLEA 9456.3 CAD
ABCA_AB315250-1ML ABCA_AB315250-100U ABCA_AB315250-20UL
Anti-Collagen IV Rabbit Recombinant Multiclonal Antibody [clone: RM1065]
Antibodies
Rabbit recombinant multiclonal [RM1065] to Collagen IV.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Collagen IV
Clonality: Recombinant Multiclonal
Clone: RM1065
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR