Anti-CD163 Rabbit Monoclonal Antibody [clone: EPR19518] (PE)

Supplier: Abcam
AB314947-10UL AB314947-100UL
ABCA_AB314947-10ULEA 366.56 CAD
ABCA_AB314947-10UL ABCA_AB314947-100U
Anti-CD163 Rabbit Monoclonal Antibody [clone: EPR19518] (PE)
Antibodies
PE Rabbit monoclonal [EPR19518] to CD163.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: EPR19518
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR