Anti-PKC delta Rabbit Monoclonal Antibody [clone: EPR17075]

Supplier: Abcam
AB182126-100UL AB182126-1ML
ABCA_AB182126-100UEA 1287.7 CAD
ABCA_AB182126-100U ABCA_AB182126-1ML
Anti-PKC delta Rabbit Monoclonal Antibody [clone: EPR17075]
Antibodies
Rabbit monoclonal [EPR17075] to PKC delta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PKC delta
Clonality: Monoclonal
Clone: EPR17075
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR