Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: EP1582Y]

Supplier: Abcam
AB62363-1ML AB62363-100UL
ABCA_AB62363-1MLEA 8901.72 CAD
ABCA_AB62363-1ML ABCA_AB62363-100UL
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: EP1582Y]
Antibodies
Rabbit monoclonal [EP1582Y] to GCDFP 15.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GCDFP 15
Clonality: Monoclonal
Clone: EP1582Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR