Anti-Heparan Sulfate Proteoglycan 2/Perlecan Rabbit Recombinant Multiclonal Antibody [clone: RM2059]

Supplier: Abcam
AB318285-1MG AB318285-100UG
ABCA_AB318285-1MGEA 16158.66 CAD
ABCA_AB318285-1MG ABCA_AB318285-100U
Anti-Heparan Sulfate Proteoglycan 2/Perlecan Rabbit Recombinant Multiclonal Antibody [clone: RM2059]
Antibodies
Rabbit recombinant multiclonal [RM2059] to Heparan Sulfate Proteoglycan 2/Perlecan - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Heparan Sulfate Proteoglycan 2/Perlecan
Clonality: Recombinant Multiclonal
Clone: RM2059
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR