Anti-PP2A alpha + beta Rabbit Monoclonal Antibody [clone: YE351]

Supplier: Abcam
AB32065-100UL AB32065-1ML
ABCA_AB32065-100ULEA 1030.16 CAD
ABCA_AB32065-100UL ABCA_AB32065-1ML
Anti-PP2A alpha + beta Rabbit Monoclonal Antibody [clone: YE351]
Antibodies
Rabbit monoclonal [YE351] to PP2A alpha + beta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PP2A alpha + beta
Clonality: Monoclonal
Clone: YE351
Conjugation:
Epitope: ab32065 reacts with an epitope located in the N terminal region of PP2A alpha.
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR