Anti-Gelsolin Mouse Monoclonal Antibody [clone: 2/Gelsolin]

Supplier: Abcam
AB303676-100UG AB303676-20UG AB303676-10UG AB303676-1MG
ABCA_AB303676-100UEA 1257.68 CAD
ABCA_AB303676-100U ABCA_AB303676-20UG ABCA_AB303676-10UG ABCA_AB303676-1MG
Anti-Gelsolin Mouse Monoclonal Antibody [clone: 2/Gelsolin]
Antibodies
Mouse monoclonal [2/Gelsolin] to Gelsolin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Gelsolin
Clonality: Monoclonal
Clone: 2/Gelsolin
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR