Anti-LDL Receptor Rabbit Monoclonal Antibody [clone: EPR24874-56]

Supplier: Abcam
AB271189-20UL AB271189-1ML AB271189-100UL
ABCA_AB271189-20ULEA 455.04 CAD
ABCA_AB271189-20UL ABCA_AB271189-1ML ABCA_AB271189-100U
Anti-LDL Receptor Rabbit Monoclonal Antibody [clone: EPR24874-56]
Antibodies
Rabbit monoclonal [EPR24874-56] to LDL Receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LDL Receptor
Clonality: Monoclonal
Clone: EPR24874-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR