Anti-Rif1 Rabbit Monoclonal Antibody [clone: EPR27131-83]

Supplier: Abcam
AB316748-1ML AB316748-10UL AB316748-100UL AB316748-20UL
ABCA_AB316748-1MLEA 11287.52 CAD
ABCA_AB316748-1ML ABCA_AB316748-10UL ABCA_AB316748-100U ABCA_AB316748-20UL
Anti-Rif1 Rabbit Monoclonal Antibody [clone: EPR27131-83]
Antibodies
Rabbit monoclonal [EPR27131-83] to Rif1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Rif1
Clonality: Monoclonal
Clone: EPR27131-83
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR