Anti-IP10 Rabbit Monoclonal Antibody [clone: EPR24674-84]

Supplier: Abcam
AB306587-10UL AB306587-100UL AB306587-20UL AB306587-1ML
ABCA_AB306587-10ULEA 396.58 CAD
ABCA_AB306587-10UL ABCA_AB306587-100U ABCA_AB306587-20UL ABCA_AB306587-1ML
Anti-IP10 Rabbit Monoclonal Antibody [clone: EPR24674-84]
Antibodies
Rabbit monoclonal [EPR24674-84] to IP10.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IP10
Clonality: Monoclonal
Clone: EPR24674-84
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR