Anti-Histone H3 (acetyl K4) Rabbit Monoclonal Antibody [clone: EPR16596]

Supplier: Abcam
AB240195-1MG AB240195-100UG
ABCA_AB240195-1MGEA 16158.66 CAD
ABCA_AB240195-1MG ABCA_AB240195-100U
Anti-Histone H3 (acetyl K4) Rabbit Monoclonal Antibody [clone: EPR16596]
Antibodies
Rabbit monoclonal [EPR16596] to Histone H3 (acetyl K4) - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR16596
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR