Anti-STAT6 Mouse Monoclonal Antibody [clone: 23/Stat6]

Supplier: Abcam
AB282107-100UG AB282107-1MG AB282107-20UG
ABCA_AB282107-100UEA 1237.14 CAD
ABCA_AB282107-100U ABCA_AB282107-1MG ABCA_AB282107-20UG
Anti-STAT6 Mouse Monoclonal Antibody [clone: 23/Stat6]
Antibodies
Mouse monoclonal [23/Stat6] to STAT6.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: EPR23939-25
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR