Anti-Topoisomerase II alpha+Topoisomerase II beta Rabbit Monoclonal Antibody [clone: EPR5377]

Supplier: Abcam
AB239984-1MG AB239984-100UG
ABCA_AB239984-1MGEA 16158.66 CAD
ABCA_AB239984-1MG ABCA_AB239984-100U
Anti-Topoisomerase II alpha+Topoisomerase II beta Rabbit Monoclonal Antibody [clone: EPR5377]
Antibodies
Rabbit monoclonal [EPR5377] to Topoisomerase II alpha + Topoisomerase II beta/TOP2B - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Topoisomerase II alpha+Topoisomerase II beta
Clonality: Monoclonal
Clone: EPR5377
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR