Anti-GABA Transporter 1 / GAT 1 Rabbit Antibody [clone: EPR12999(2)]

Supplier: Abcam
AB185205-100UL AB185205-1ML
ABCA_AB185205-100UEA 1060.18 CAD
ABCA_AB185205-100U ABCA_AB185205-1ML
Anti-GABA Transporter 1 / GAT 1 Rabbit Antibody [clone: EPR12999(2)]
Antibodies
Rabbit monoclonal [EPR12999(2)] to GABA Transporter 1 / GAT 1.

Type: Primary
Antigen: GABA Transporter 1 / GAT 1
Clonality:
Clone: EPR12999(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR