Carboxyl-Gold, nanoparticles, 40 nm
Supplier: Abcam
AB269942-40UG
AB269942-200UG
ABCA_AB269942-40UGEA
1039.64
CAD
ABCA_AB269942-40UG
ABCA_AB269942-200U
Carboxyl-Gold, nanoparticles, 40 nm
Gold
Carboxyl Gold Nanoparticles (40nm, 40OD) are coated gold nanoparticles functionalized with carboxyl groups that allow the covalent conjugation of antibodies, using the water-soluble carbodiimide crosslinker EDC.