Anti-MICA+MICB Rabbit Monoclonal Antibody [clone: EPR24086-121] (APC (Allophycocyanin))

Supplier: Abcam
AB314409-100UL AB314409-10UL
ABCA_AB314409-100UEA 1248.2 CAD
ABCA_AB314409-100U ABCA_AB314409-10UL
Anti-MICA+MICB Rabbit Monoclonal Antibody [clone: EPR24086-121] (APC (Allophycocyanin))
Antibodies
APC Rabbit monoclonal [EPR24086-121] to MICA + MICB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MICA+MICB
Clonality: Monoclonal
Clone: EPR24086-121
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR