Anti-Axl Mouse Monoclonal Antibody [clone: 70000000000] (PE)

Supplier: Abcam
AB313946-10UL AB313946-100UL
ABCA_AB313946-10ULEA 366.56 CAD
ABCA_AB313946-10UL ABCA_AB313946-100U
Anti-Axl Mouse Monoclonal Antibody [clone: 70000000000] (PE)
Antibodies
PE Mouse monoclonal [7E10] to Axl.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Axl
Clonality: Monoclonal
Clone: 70000000000
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR